CAS RN 94724-12-6
IUPAC Name:
[2-(4-methoxy-3-{[2-methoxy-3-({2-methoxy-5-[2-(methylamino)ethyl]phenyl}methyl)-5-[2-(methylamino)ethyl]phenyl]methyl}phenyl)ethyl](methyl)amine trihydrochloride
Synonyms:
[2-(4-methoxy-3-{[2-methoxy-3-({2-methoxy-5-[2-(methylamino)ethyl]phenyl}methyl)-5-[2-(methylamino)ethyl]phenyl]methyl}phenyl)ethyl](methyl)amine trihydrochloride
Molecular Weight (g/mol):
629.1
Molecular Formula:
C32H48Cl3N3O3
InChi Key:
KIAWAQJORBNOIF-UHFFFAOYSA-N
SMILES:
Cl.Cl.Cl.CNCCC1=CC(CC2=CC(CCNC)=CC(CC3=C(OC)C=CC(CCNC)=C3)=C2OC)=C(OC)C=C1
1
–
1
of
1
results
1
Compound 48/80, MP Biomedicals™
CAS: 94724-12-6 Molecular Formula: C32H48Cl3N3O3 Molecular Weight (g/mol): 629.104 InChI Key: KIAWAQJORBNOIF-UHFFFAOYSA-N Synonym: compound 48/80 trihydrochloride PubChem CID: 118797323 IUPAC Name: 2-[4-methoxy-3-[[2-methoxy-3-[[2-methoxy-5-[2-(methylamino)ethyl]phenyl]methyl]-5-[2-(methylamino)ethyl]phenyl]methyl]phenyl]-N-methylethanamine;trihydrochloride SMILES: CNCCC1=CC(=C(C=C1)OC)CC2=CC(=CC(=C2OC)CC3=C(C=CC(=C3)CCNC)OC)CCNC.Cl.Cl.Cl