Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Compound 48/80, MP Biomedicals™

Catalog No. ICN15902650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15902650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Alternative option to part number C2313
Catalog No. ICN15902650 Supplier MP Biomedicals, Inc Supplier No. 0215902650
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alternative option to part number C2313

Chemical Identifiers

CAS 94724-12-6
Molecular Formula C32H48Cl3N3O3
Molecular Weight (g/mol) 629.104
InChI Key KIAWAQJORBNOIF-UHFFFAOYSA-N
Synonym compound 48/80 trihydrochloride
PubChem CID 118797323
IUPAC Name 2-[4-methoxy-3-[[2-methoxy-3-[[2-methoxy-5-[2-(methylamino)ethyl]phenyl]methyl]-5-[2-(methylamino)ethyl]phenyl]methyl]phenyl]-N-methylethanamine;trihydrochloride
SMILES CNCCC1=CC(=C(C=C1)OC)CC2=CC(=CC(=C2OC)CC3=C(C=CC(=C3)CCNC)OC)CCNC.Cl.Cl.Cl

Specifications

Color White
Quantity 50 mg
Formula Weight 630
Physical Form Powder
Chemical Name or Material Compound 48/80
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.