accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 947533-23-5

IUPAC Name: [4-(cyanomethoxy)phenyl]boronic acid
Synonyms: 4-(cyanomethoxy)phenylboronic acid
Molecular Weight (g/mol): 176.97
Molecular Formula: C8H8BNO3
InChi Key: GKJPNYHKHSLHCC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(OCC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results