Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Cyanomethoxy)phenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog No. AC445400010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445400010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC445400010 Supplier Thermo Scientific Chemicals Supplier No. 445400010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 947533-23-5 | Molecular Formula: C8H8BNO3 | Molecular Weight: 176.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947533-23-5
Molecular Formula C8H8BNO3
Molecular Weight (g/mol) 176.97
MDL Number MFCD09908170
InChI Key GKJPNYHKHSLHCC-UHFFFAOYSA-N
Synonym 4-cyanomethoxyphenylboronic acid, 4-cyanomethoxy phenyl boronic acid, 4-cyanomethoxy phenylboronic acid, acmc-209rtj
PubChem CID 21918429
IUPAC Name [4-(cyanomethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=C(OCC#N)C=C1

Specifications

Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 176.97
Percent Purity 97%
Chemical Name or Material 4-(Cyanomethoxy)phenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.