accessibility menu, dialog, popup

UserName

Viewing profile as user:
BONHOOH

CAS RN 947533-25-7

IUPAC Name: [3-(cyanomethoxy)phenyl]boronic acid
Synonyms: 3-(cyanomethoxy)phenylboronic acid
Molecular Weight (g/mol): 176.97
Molecular Formula: C8H8BNO3
InChi Key: QEXGRJBUDVWWSC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(OCC#N)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results