accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 948-03-8

IUPAC Name: methyl 1-hydroxynaphthalene-2-carboxylate
Synonyms: methyl 1-hydroxynaphthalene-2-carboxylate
Molecular Weight (g/mol): 202.21
Molecular Formula: C12H10O3
InChi Key: HMIBDRSTVGFJPB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2C=CC=CC2=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)