Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-hydroxy-2-naphthoate

Catalog No. AC428470050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428470050 5 g Glass bottle
AC428470010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428470050 Supplier Thermo Scientific Chemicals Supplier No. 428470050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 948-03-8 | C12H10O3 | 202.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 948-03-8
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.21
InChI Key HMIBDRSTVGFJPB-UHFFFAOYSA-N
Synonym methyl 1-hydroxy-2-naphthoate, 2-naphthalenecarboxylic acid, 1-hydroxy-, methyl ester, 1-hydroxy-2-naphthoic acid methyl ester, 1-hydroxy-2-naphthoic acid, methyl ester, acmc-20akjf, 2-naphthoic acid, 1-hydroxy-, methyl ester, methyl=1-hydroxy-2-naphthoate, 1-hydroxy-2-carbomethoxynaphthalene, methyl 1-hydroxy-naphthalene-2-carboxylate
PubChem CID 70358
IUPAC Name methyl 1-hydroxynaphthalene-2-carboxylate
SMILES COC(=O)C1=C(C2=CC=CC=C2C=C1)O

Specifications

Melting Point 76°C to 80°C
Packaging Glass bottle
Quantity 5 g
Formula Weight 202.21
Chemical Name or Material Methyl 1-hydroxy-2-naphthoate
Hazard Category GHS Signal Word: Warning
EINECSNumber 213-434-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.