accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(S)NH2HO

CAS RN 949-67-7

IUPAC Name: ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Synonyms: L-tyrosine, ethyl ester Tyr-OEt
Molecular Weight (g/mol): 209.25
Molecular Formula: C11H15NO3
InChi Key: SBBWEQLNKVHYCX-JTQLQIEISA-N
SMILES: CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results