Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Tyrosine Ethyl Ester 98.0+%, TCI America™
SDP

Catalog No. T05515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T05515G 5 g
T05511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T05515G Supplier TCI America Supplier No. T05515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 949-67-7
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00063046
InChI Key SBBWEQLNKVHYCX-JTQLQIEISA-N
Synonym l-tyrosine ethyl ester, ethyl l-tyrosinate, tyrosine ethyl ester, h-tyr-oet, l-tyrosine, ethyl ester, tyr-oet, ethyl tyrosine ester, l-tyrosine ethyl, unii-17rpw76a9g, s-ethyl 2-amino-3-4-hydroxyphenyl propanoate
PubChem CID 70364
ChEBI CHEBI:9798
IUPAC Name ethyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES CCOC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 5 g
Formula Weight 209.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Tyrosine Ethyl Ester
EINECSNumber (3)-1580
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.