accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNCH3(E/Z)

CAS RN 94941-30-7

IUPAC Name: 1-(9-ethyl-9H-carbazol-3-yl)-N-(2-methyl-2,3-dihydro-1H-indol-1-yl)methanimine
Synonyms: 1-(9-ethylcarbazol-3-yl)-N-(2-methyl-2,3-dihydroindol-1-yl)methanimine
Molecular Weight (g/mol): 353.47
Molecular Formula: C24H23N3
InChi Key: TWPXERWDHZWUPC-UHFFFAOYNA-N
SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC(C=NN1C(C)CC3=CC=CC=C13)=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results