Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(9-Ethylcarbazol-3-yl)methylene]-2-methyl-1-indolinylamine, TCI America™
SDP

Catalog No. E05721G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E05721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E05721G Supplier TCI America Supplier No. E05721G

Chemical Identifiers

CAS 94941-30-7
Molecular Formula C24H23N3
Molecular Weight (g/mol) 353.469
MDL Number MFCD06797100
InChI Key TWPXERWDHZWUPC-XYGWBWBKSA-N
Synonym 9-Ethyl-3-[[(2-methyl-1-indolinyl)imino]methyl]carbazole
PubChem CID 67045615
IUPAC Name (Z)-1-(9-ethylcarbazol-3-yl)-N-(2-methyl-2,3-dihydroindol-1-yl)methanimine
SMILES CCN1C2=C(C=C(C=C2)C=NN3C(CC4=CC=CC=C43)C)C5=CC=CC=C51

Specifications

Color Yellow
Quantity 1 g
Formula Weight 353.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-[(9-Ethylcarbazol-3-yl)methylene]-2-methyl-1-indolinylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.