accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95-10-3

IUPAC Name: 1-{bicyclo[2.2.1]hept-5-en-2-yl}methanamine
Synonyms: 1-{bicyclo[2.2.1]hept-5-en-2-yl}methanamine
Molecular Weight (g/mol): 123.2
Molecular Formula: C8H13N
InChi Key: XLBALIGLOMYEKN-UHFFFAOYNA-N
SMILES: NCC1CC2CC1C=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results