Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Norbornene-2-methylamine (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. N090725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N090725G 25 g
N09075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N090725G Supplier TCI America Supplier No. N090725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 95-10-3
Molecular Formula C8H13N
Molecular Weight (g/mol) 123.199
MDL Number MFCD07369043
InChI Key XLBALIGLOMYEKN-UHFFFAOYSA-N
Synonym 5-(Aminomethyl)bicyclo[2.2.1]hept-2-ene, 5-(Aminomethyl)norbornene, 5-Norbornene-2-methanamine
PubChem CID 247182
IUPAC Name 5-bicyclo[2.2.1]hept-2-enylmethanamine
SMILES C1C2CC(C1C=C2)CN

Specifications

Color Yellow
Quantity 25 g
UN Number 2735
Formula Weight 123.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Norbornene-2-methylamine (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.