accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH

CAS RN 95-12-5

IUPAC Name: {bicyclo[2.2.1]hept-5-en-2-yl}methanol
Synonyms: bicyclo[2.2.1]hept-5-en-2-ylmethanol
Molecular Weight (g/mol): 124.18
Molecular Formula: C8H12O
InChi Key: LUMNWCHHXDUKFI-UHFFFAOYNA-N
SMILES: OCC1CC2CC1C=C2
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)