Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo(2.2.1)hept-5-ene-2-methanol, MP Biomedicals

Catalog No. ICN214388 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214388 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214388 Supplier MP Biomedicals, Inc Supplier No. 0521438805

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 95-12-5
Molecular Formula C8H12O
Molecular Weight (g/mol) 124.183
InChI Key LUMNWCHHXDUKFI-UHFFFAOYSA-N
Synonym 5-norbornene-2-methanol, bicyclo 2.2.1 hept-5-en-2-ylmethanol, bicyclo 2.2.1 hept-5-ene-2-methanol, cyclol, 2-hydroxymethyl-5-norbornene, 5-hydroxymethyl-2-norbornene, 5-norbornene 2-methanol, 5-hydroxymethylbicyclo 2.2.1 hept-2-ene, 2-hydroxymethyl bicyclo 2.2.1 hept-5-ene
PubChem CID 78946
IUPAC Name 5-bicyclo[2.2.1]hept-2-enylmethanol
SMILES C1C2CC(C1C=C2)CO

Specifications

Quantity 5 g
Chemical Name or Material Bicyclo(2.2.1)hept-5-ene-2-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.