accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95-29-4

IUPAC Name: (1,3-benzothiazol-2-ylsulfanyl)bis(propan-2-yl)amine
Synonyms: (1,3-benzothiazol-2-ylsulfanyl)diisopropylamine
Molecular Weight (g/mol): 266.42
Molecular Formula: C13H18N2S2
InChi Key: ILSQBBRAYMWZLQ-UHFFFAOYSA-N
SMILES: CC(C)N(SC1=NC2=CC=CC=C2S1)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results