Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diisopropylbenzothiazole-2-sulfenamide, MP Biomedicals

Catalog No. ICN207047 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN207047 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN207047 Supplier MP Biomedicals, Inc Supplier No. 0520704780

Chemical Identifiers

CAS 95-29-4
Molecular Formula C13H18N2S2
Molecular Weight (g/mol) 266.421
InChI Key ILSQBBRAYMWZLQ-UHFFFAOYSA-N
Synonym meramid p, n,n-diisopropylbenzothiazole-2-sulfenamide, dibs, dipac, santocure ips, dipac accelerator, dipak, 2-benzothiazolesulfenamide, n,n-bis 1-methylethyl, unii-3lh1p5muk0, n,n-diisopropylbenzothiazole-2-sulphenamide
PubChem CID 7228
IUPAC Name N-(1,3-benzothiazol-2-ylsulfanyl)-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)SC1=NC2=CC=CC=C2S1

Specifications

Quantity 100 g
Formula Weight 282.45
Chemical Name or Material N, N-Diisopropylbenzothiazole-2-sulfenamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.