accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 95-32-9

IUPAC Name: 2-(morpholin-4-yldisulfanyl)-1,3-benzothiazole
Synonyms: 2-(morpholin-4-yldisulfanyl)-1,3-benzothiazole
Molecular Weight (g/mol): 284.41
Molecular Formula: C11H12N2OS3
InChi Key: QRYFCNPYGUORTK-UHFFFAOYSA-N
SMILES: C1CN(CCO1)SSC1=NC2=CC=CC=C2S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results