Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Benzothiazolyldithio)morpholine 95.0+%, TCI America™
SDP

Catalog No. B134925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B134925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B134925G Supplier TCI America Supplier No. B134925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95-32-9
Molecular Formula C11H12N2OS3
Molecular Weight (g/mol) 284.41
MDL Number MFCD00059033
InChI Key QRYFCNPYGUORTK-UHFFFAOYSA-N
PubChem CID 7231
IUPAC Name 2-(morpholin-4-yldisulfanyl)-1,3-benzothiazole
SMILES C1CN(CCO1)SSC1=NC2=CC=CC=C2S1

Specifications

Melting Point 130°C
Color Yellow
Quantity 25 g
UN Number 3077
Formula Weight 284.41
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2-Benzothiazolyldithio)morpholine
RTECSNumber DL5953000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.