CAS RN 95-45-4
Dimethylglyoxime, 99+%
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime, 99%
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 IUPAC Name: N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime, ACS, 99+%
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 IUPAC Name: N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime, 99+%, ACS reagent
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime, ACS Reagent Grade, Ricca Chemical
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 IUPAC Name: (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime 98.0+%, TCI America™
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 IUPAC Name: (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime RS, 1% (w/v) Alcoholic Solution, Ricca Chemical
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 135399895 IUPAC Name: (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime, ACS Grade, LabChem™
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 IUPAC Name: (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime, 1% Alcoholic, for Nickel, Certified, LabChem™
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 135399895 IUPAC Name: (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O
Sigma Aldrich Dimethylglyoxime
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Dimethylglyoxime
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Dimethylglyoxime
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fisher Science Education™ Dimethylglyoxime
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 IUPAC Name: (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime (Crystalline./Certified ACS), Fisher Chemical
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 IUPAC Name: (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O
Dimethylglyoxime, For ACS analysis, 99.49%, MP Biomedicals™
CAS: 95-45-4 Molecular Formula: C4H8N2O2 Molecular Weight (g/mol): 116.12 MDL Number: MFCD00002117 InChI Key: JGUQDUKBUKFFRO-GGWOSOGESA-N Synonym: biacetyl dioxime,dimethylglyoxime,diacetyldioxime,2,3-butanedione dioxime,diacetyl dioxime,2,3-diisonitrosobutane,biacetyl, dioxime,chugaev's reagent,nsc 9,unii-2971mft1ky PubChem CID: 5356010 IUPAC Name: (E)-N-[(3E)-3-(hydroxyimino)butan-2-ylidene]hydroxylamine SMILES: C\C(=N/O)\C(\C)=N\O