Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethylglyoxime

Catalog No. 03858500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-858-500G 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-858-500G Supplier Sigma Aldrich Supplier No. 03858500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N,n-dimethyldecylamine is a tertiary amine. it can be prepared by reacting n,n-dimethyldodecylamine, t-butyl hydroperoxide, vanadium oxyacetylacetonate and t-butyl alcohol.n,n-dimethyldecylamine may be used as one of the reaction components in the preparation of:
  • Comb-shaped quaternized poly(2,6-dimethylphenylene oxide) (ppo)
  • Pore-expanded mcm-41 silica
  • Qa c10 stationary phase bearing quaternary ammonium embedded between a propyl and a decyl chain

Specifications

CAS 95-45-4
Melting Point 240°C to 241°C (lit.)
Molecular Formula C4H8N2O2
Linear Formula CH3C(=NOH)C(=NOH)CH3
MDL Number MFCD00002117
Quantity 1 kg
Synonym dmgH2; 2,3-Butanedione dioxime; Diacetyldioxime
Molecular Weight (g/mol) 116.12
Percent Purity ≥97.0% (TLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.