CAS RN 95-48-7
o-Cresol, 99%, AcroSeal™
CAS: 95-48-7 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 MDL Number: MFCD00002226 InChI Key: QWVGKYWNOKOFNN-UHFFFAOYSA-N Synonym: o-cresol,2-hydroxytoluene,o-methylphenol,2-cresol,phenol, 2-methyl,orthocresol,o-cresylic acid,o-oxytoluene,o-toluol,o-hydroxytoluene PubChem CID: 335 ChEBI: CHEBI:28054 IUPAC Name: 2-methylphenol SMILES: CC1=CC=CC=C1O
o-Cresol, 98+%
CAS: 95-48-7 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 MDL Number: MFCD00002226 InChI Key: QWVGKYWNOKOFNN-UHFFFAOYSA-N Synonym: o-cresol,2-hydroxytoluene,o-methylphenol,2-cresol,phenol, 2-methyl,orthocresol,o-cresylic acid,o-oxytoluene,o-toluol,o-hydroxytoluene PubChem CID: 335 ChEBI: CHEBI:28054 IUPAC Name: 2-methylphenol SMILES: CC1=CC=CC=C1O
o-Cresol, 99%
CAS: 95-48-7 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 MDL Number: MFCD00002226 InChI Key: QWVGKYWNOKOFNN-UHFFFAOYSA-N Synonym: o-cresol,2-hydroxytoluene,o-methylphenol,2-cresol,phenol, 2-methyl,orthocresol,o-cresylic acid,o-oxytoluene,o-toluol,o-hydroxytoluene PubChem CID: 335 ChEBI: CHEBI:28054 IUPAC Name: 2-methylphenol SMILES: CC1=CC=CC=C1O
o-Cresol 99.0+%, TCI America™
CAS: 95-48-7 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 MDL Number: MFCD00002226 InChI Key: QWVGKYWNOKOFNN-UHFFFAOYSA-N Synonym: o-cresol,2-hydroxytoluene,o-methylphenol,2-cresol,phenol, 2-methyl,orthocresol,o-cresylic acid,o-oxytoluene,o-toluol,o-hydroxytoluene PubChem CID: 335 ChEBI: CHEBI:28054 IUPAC Name: 2-methylphenol SMILES: CC1=CC=CC=C1O
2-Methylphenol, SPEX CertiPrep™
CAS: 95-48-7 Molecular Formula: C7H8O Molecular Weight (g/mol): 108.14 MDL Number: MFCD00002226 InChI Key: QWVGKYWNOKOFNN-UHFFFAOYSA-N PubChem CID: 335 ChEBI: CHEBI:28054 IUPAC Name: 2-methylphenol SMILES: CC1=CC=CC=C1O