Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Cresol (Laboratory), Fisher Chemical

Catalog No. C536500 Shop All Fisherbrand Products
Change view
Click to view available options
Glass Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
C536-500 Glass Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C536-500

missing translation for 'validMainframeMsg'

Specifications

CAS 95-48-7
Melting Point 30°C
Color Amber
Density 1.040g/cm³
pH 4.8
Boiling Point 191°C
Packaging Glass Bottle
Molecular Formula C7H8O
Linear Formula CH3C6H4OH
Quantity 500 g
MDL Number MFCD00002226
Synonym o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
InChI Key QWVGKYWNOKOFNN-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1O
IUPAC Name 2-methylphenol
Molecular Weight (g/mol) 108.14
PubChem CID 335
ChEBI CHEBI:28054
Vapor Pressure 0.168mmHg at 20°C
Formula Weight 108.14
Grade Laboratory
Physical Form Solid
Chemical Name or Material o-Cresol
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Combustible material. Causes burns by all exposure routes. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:2
Instability:1
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.