accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3Cl

CAS RN 95-49-8

IUPAC Name: 1-chloro-2-methylbenzene
Synonyms: 2-chlorotoluene
Molecular Weight (g/mol): 126.58
Molecular Formula: C7H7Cl
InChi Key: IBSQPLPBRSHTTG-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1Cl
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)