Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorotoluene Solution, 5000μg/mL in Methanol, Ultra Scientific

Catalog No. USEPA1018
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1018 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1018 Supplier Agilent Technologies Supplier No. EPA10181
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95-49-8 , 67-56-1
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000562
InChI Key IBSQPLPBRSHTTG-UHFFFAOYSA-N
PubChem CID 7238
IUPAC Name 1-chloro-2-methylbenzene
SMILES CC1=CC=CC=C1Cl

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material 2-Chlorotoluene, 5000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.