CAS RN 95-50-1
1,2-Dichlorobenzene, HPLC Grade, 98% min
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 146.998 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, 99%, pure
CAS: 95-50-1 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, 99+%, for HPLC
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 147 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene 99.0+%, TCI America™
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 146.998 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, 99%
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 146.998 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, 98+%, Extra Dry, AcroSeal™
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 147 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, Spectrophotometric Grade, 98%
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 146.998 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
1,2-Dichlorobenzene, 99+%, for spectroscopy
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 147 MDL Number: MFCD00000535 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N Synonym: o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl
Fluoride/Chloride in Oil - PT, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
1,2-Dichlorobenzene, SPEX CertiPrep™
CAS: 95-50-1 Molecular Formula: C6H4Cl2 Molecular Weight (g/mol): 146.998 InChI Key: RFFLAFLAYFXFSW-UHFFFAOYSA-N PubChem CID: 7239 ChEBI: CHEBI:35290 IUPAC Name: 1,2-dichlorobenzene SMILES: C1=CC=C(C(=C1)Cl)Cl