Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichlorobenzene, 98% min., MilliporeSigma™

Catalog No. MDX06853 Shop All MilliporeSigma Products
Change view
Click to view available options
4L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MDX06853 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MDX06853 Supplier MilliporeSigma Supplier No. DX06853

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

Specifications

CAS 95-50-1
Color Colorless
Density 1.31g/cm3 (20°C)
Explosion Limit 2.2 to 12%(V)
Ignition Point 640°C
Assay Percent Range 98% min.
Molecular Formula C6H4Cl2
Linear Formula 1, 2-(Cl)2C6H4
Quantity 4 L
Synonym o-dichlorobenzene, chloroben, o-dichlorbenzol, dilantin db, ortho-dichlorobenzene, 2-dichlorobenzene, o-dichlorobenzol, odcb, benzene, 1,2-dichloro, cloroben
InChI Key RFFLAFLAYFXFSW-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)Cl)Cl
IUPAC Name 1,2-dichlorobenzene
Molecular Weight (g/mol) 146.998
PubChem CID 7239
ChEBI CHEBI:35290
Formula Weight 147
Vapor Pressure 1.33 hPa (20°C)
Physical Form Liquid
Chemical Name or Material 1, 2-Dichlorobenzene
Show More Show Less
Recommended Storage Store at +15°C to +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.