accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCl

CAS RN 95-58-9

IUPAC Name: 2-chloro-3-methylpyrazine
Synonyms: 2-chloro-3-methylpyrazine
Molecular Weight (g/mol): 128.56
Molecular Formula: C5H5ClN2
InChi Key: WZHWPZQQPWKEAV-UHFFFAOYSA-N
SMILES: CC1=NC=CN=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)