CAS RN 950201-71-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Fluoro-4-(4-tetrahydropyranylamino)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 950201-71-5 Synonym: 3-Fluoro-4-(4-tetrahydropyranylamino)phenylboronic acid pinacol ester; [2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-phenyl](tetrahydropyran-4-yl)-amine