accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBrOBrBrBrNOO

CAS RN 95111-49-2

IUPAC Name: 2,3,5,6-tetrabromo-4-methyl-4-nitrocyclohexa-2,5-dien-1-one
Synonyms: 2,3,5,6-tetrabromo-4-methyl-4-nitrocyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 468.72
Molecular Formula: C7H3Br4NO3
InChi Key: KKWDBDVGMIDKLP-UHFFFAOYSA-N
SMILES: CC1(C(Br)=C(Br)C(=O)C(Br)=C1Br)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)