Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrabromo-4-methyl-4-nitrocyclohexa-2,5-dienone 97%, Thermo Scientific™

Catalog No. AC257680010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC257680010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC257680010 Supplier Thermo Scientific Chemicals Supplier No. 257680010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95111-49-2
Molecular Formula C7H3Br4NO3
Molecular Weight (g/mol) 468.721
InChI Key KKWDBDVGMIDKLP-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrabromo-4-methyl-4-nitro-2,5-cyclohexadien-1-one, acmc-209ruw, 2,3,5,6-tetrabromo-4-methyl-4-nitrocyclohexa-2,5-dienone, 4-nitro-4-methyl-2,3,5,6-tetrabromo-2,5-cyclohexadien-1-one, 2,3,5,6-tetrabromo-4-methyl-4-nitro-cyclohexa-2,5-dien-1-one
PubChem CID 2724547
IUPAC Name 2,3,5,6-tetrabromo-4-methyl-4-nitrocyclohexa-2,5-dien-1-one
SMILES CC1(C(=C(C(=O)C(=C1Br)Br)Br)Br)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 88°C to 93°C
Color Beige-Yellow to Brown
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 468.71
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrabromo-4-methyl-4-nitrocyclohexa-2,5-dienone, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.