accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SMgBr

CAS RN 95184-07-9

IUPAC Name: 2-(bromomagnesio)-3-methylthiophene
Synonyms: 2-(bromomagnesio)-3-methylthiophene
Molecular Weight (g/mol): 201.36
Molecular Formula: C5H5BrMgS
InChi Key: OVSYYYAKOISBKJ-UHFFFAOYSA-M
SMILES: CC1=C([Mg]Br)SC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results