accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 952-47-6

IUPAC Name: (6E)-4-methyl-6-(2-phenylhydrazin-1-ylidene)cyclohexa-2,4-dien-1-one
Synonyms: (6E)-4-methyl-6-(2-phenylhydrazin-1-ylidene)cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: YCUUKIHFSTZZIS-NTCAYCPXSA-N
SMILES: CC1=C\C(=N/NC2=CC=CC=C2)C(=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results