Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylazo-4-methylphenol 98.0+%, TCI America™
SDP

Catalog No. P014525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P014525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P014525G Supplier TCI America Supplier No. P014525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 952-47-6
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00045788
InChI Key YCUUKIHFSTZZIS-NTCAYCPXSA-N
Synonym 2-Hydroxy-5-methylazobenzene, 4-Methyl-2-phenylazophenol, 2-Phenylazo-p-cresol
PubChem CID 5370037
IUPAC Name (6E)-4-methyl-6-(2-phenylhydrazin-1-ylidene)cyclohexa-2,4-dien-1-one
SMILES CC1=C\C(=N/NC2=CC=CC=C2)C(=O)C=C1

Specifications

Melting Point 105°C
Quantity 25 g
Formula Weight 212.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylazo-4-methylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.