accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOOHHCl

CAS RN 953079-94-2

IUPAC Name: pyrrolidine-3-carboxylic acid hydrochloride
Synonyms: pyrrolidine-3-carboxylic acid hydrochloride
Molecular Weight (g/mol): 151.59
Molecular Formula: C5H10ClNO2
InChi Key: OYCLYMMIZJWYJG-UHFFFAOYNA-N
SMILES: Cl.OC(=O)C1CCNC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)