Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-beta-Proline hydrochloride, 95%

Catalog No. AAH57365MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57365MD 250 mg
AAH5736503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57365MD Supplier Thermo Scientific Chemicals Supplier No. H57365MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 953079-94-2 | C5H10ClNO2 | 151.59 g/mol

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is used as a key intermediate to prepare two different pyrrolidines.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 953079-94-2
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD06801272
InChI Key OYCLYMMIZJWYJG-UHFFFAOYSA-N
Synonym pyrrolidine-3-carboxylic acid hydrochloride, beta-proline hcl, 3-pyrrolidinecarboxylic acid hcl, 3-pyrrolidinecarboxylic acid hydrochloride, pubchem16223, beta-proline hydrochloride, ksc494m2f, pyrrolidine-3-carboxylicacidhydrochloride, 3-pyrrolidine carboxylic acid hydrochloride, rs-pyrrolidine-3-carboxylic acid hydrochloride
PubChem CID 44890816
IUPAC Name pyrrolidine-3-carboxylic acid;hydrochloride
SMILES C1CNCC1C(=O)O.Cl

Specifications

Melting Point 30°C to 38°C
Quantity 250 mg
Flash Point >110°C (230°F)
Formula Weight 151.59
Percent Purity 95%
Chemical Name or Material DL-beta-Proline hydrochloride
Hazard Category H318
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement P280-P305+P351+P338-P310
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.