accessibility menu, dialog, popup

UserName

Viewing profile as user:
PPPdClClFet-Butylt-Butylt-Butylt-Butyl

CAS RN 95408-45-0

IUPAC Name: Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
Synonyms:
Molecular Weight (g/mol): 651.75
Molecular Formula: C26H44Cl2FeP2Pd
InChi Key:
SMILES: [Fe].Cl[Pd]Cl.CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)