CAS RN 95453-58-0
IUPAC Name:
2-chloro-1,3-thiazole-5-carbaldehyde
Synonyms:
2-chloro-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol):
147.58
Molecular Formula:
C4H2ClNOS
InChi Key:
PKCBQQXHFIDIIG-UHFFFAOYSA-N
SMILES:
ClC1=NC=C(S1)C=O
1
–
1
of
1
results
1
Sigma Aldrich 2-Chlorothiazole-5-carboxaldehyde
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.