accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 955977-50-1

IUPAC Name: 4-(6-{[4-(cyclopropylmethyl)piperazin-1-yl]methyl}-4-(morpholin-4-yl)thieno[3,2-d]pyrimidin-2-yl)-5-fluoro-1H-indole
Synonyms: 4-(6-{[4-(cyclopropylmethyl)piperazin-1-yl]methyl}-4-(morpholin-4-yl)thieno[3,2-d]pyrimidin-2-yl)-5-fluoro-1H-indole
Molecular Weight (g/mol): 506.64
Molecular Formula: C27H31FN6OS
InChi Key: YDNOHCOYQVZOMC-UHFFFAOYSA-N
SMILES: FC1=CC=C2NC=CC2=C1C1=NC(N2CCOCC2)=C2SC(CN3CCN(CC4CC4)CC3)=CC2=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results