accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CClO(E)

CAS RN 95636-02-5

IUPAC Name: (6E)-8-methylnon-6-enoyl chloride
Synonyms: (6E)-8-methylnon-6-enoyl chloride
Molecular Weight (g/mol): 188.7
Molecular Formula: C10H17ClO
InChi Key: JDZRIGWDAPNYLT-FNORWQNLSA-N
SMILES: CC(C)\C=C\CCCCC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results