Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-8-Methyl-6-nonenoyl Chloride 95.0+%, TCI America™
SDP

Catalog No. M18261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18261G Supplier TCI America Supplier No. M18261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95636-02-5
Molecular Formula C10H17ClO
Molecular Weight (g/mol) 188.70
MDL Number MFCD06797318
InChI Key JDZRIGWDAPNYLT-FNORWQNLSA-N
PubChem CID 6442332
IUPAC Name (6E)-8-methylnon-6-enoyl chloride
SMILES CC(C)\C=C\CCCCC(Cl)=O

Specifications

Color Yellow
Quantity 1 g
UN Number 3265
Formula Weight 188.70
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-8-Methyl-6-nonenoyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.