accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 956700-19-9

IUPAC Name: [2-({2-[bis(propan-2-yl)amino]ethyl}amino)ethyl]bis(propan-2-yl)amine
Synonyms: (2-{[2-(diisopropylamino)ethyl]amino}ethyl)diisopropylamine
Molecular Weight (g/mol): 271.49
Molecular Formula: C16H37N3
InChi Key: DVVORTHDQVBZOO-UHFFFAOYSA-N
SMILES: CC(C)N(CCNCCN(C(C)C)C(C)C)C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results