Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N″,N″-Tetraisopropyldiethylenetriamine 98.0+%, TCI America™
SDP

Catalog No. T22575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T22575G Supplier TCI America Supplier No. T22575G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 956700-19-9
Molecular Formula C16H37N3
Molecular Weight (g/mol) 271.49
MDL Number MFCD00432577
InChI Key DVVORTHDQVBZOO-UHFFFAOYSA-N
Synonym Bis[2-(diisopropylamino)ethyl]amine
PubChem CID 17793888
IUPAC Name [2-({2-[bis(propan-2-yl)amino]ethyl}amino)ethyl]bis(propan-2-yl)amine
SMILES CC(C)N(CCNCCN(C(C)C)C(C)C)C(C)C

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 271.49
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N,N′′,N′′-Tetraisopropyldiethylenetriamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.