accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSOONNSNNNHNNO

CAS RN 957054-30-7

IUPAC Name: 4-{6-[(4-methanesulfonylpiperazin-1-yl)methyl]-4-(morpholin-4-yl)thieno[3,2-d]pyrimidin-2-yl}-1H-indazole
Synonyms: 4-{6-[(4-methanesulfonylpiperazin-1-yl)methyl]-4-(morpholin-4-yl)thieno[3,2-d]pyrimidin-2-yl}-1H-indazole
Molecular Weight (g/mol): 513.64
Molecular Formula: C23H27N7O3S2
InChi Key: LHNIIDJUOCFXAP-UHFFFAOYSA-N
SMILES: CS(=O)(=O)N1CCN(CC2=CC3=NC(=NC(N4CCOCC4)=C3S2)C2=C3C=NNC3=CC=C2)CC1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results