CAS RN 957060-88-7
IUPAC Name:
(3-chloro-2-methoxypyridin-4-yl)boronic acid
Synonyms:
3-chloro-2-methoxypyridin-4-ylboronic acid
Molecular Weight (g/mol):
187.39
Molecular Formula:
C6H7BClNO3
InChi Key:
YQNAXOGPSUVHNU-UHFFFAOYSA-N
SMILES:
COC1=NC=CC(B(O)O)=C1Cl
1
–
1
of
1
results
1
Promotions Available
3-Chloro-2-methoxypyridine-4-boronic acid, 95%, Thermo Scientific™
3-Chloro-2-methoxypyridine-4-boronic acid, 95%, Thermo Scientific™
CAS: 957060-88-7 Molecular Formula: C6H7BClNO3 Molecular Weight (g/mol): 187.39 MDL Number: MFCD06798255 InChI Key: YQNAXOGPSUVHNU-UHFFFAOYSA-N Synonym: 3-chloro-2-methoxypyridine-4-boronic acid,3-chloro-2-methoxypyridin-4-yl boronic acid,3-chloro-2-methoxypyridin-4-ylboronicacid,2-methoxy-3-chloropyridine-4-boronic acid,3-chloro-2-methoxy-4-pyridineboronic acid,pubchem17058,abbypharma ap-17-5909,3-chloro-2-methoxypyrdine-4-boronic acid,boronic acid, b-3-chloro-2-methoxy-4-pyridinyl PubChem CID: 44119809 IUPAC Name: (3-chloro-2-methoxypyridin-4-yl)boronic acid SMILES: COC1=NC=CC(B(O)O)=C1Cl