CAS RN 957198-31-1
IUPAC Name:
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine
Synonyms:
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine
Molecular Weight (g/mol):
289.19
Molecular Formula:
C15H24BN3O2
InChi Key:
LOZSSHYLDBDYSQ-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC(=NC=C1)N1CCNCC1
1
–
1
of
1
results
1
Sigma Aldrich 4-Amino-3-Nitrophenylboronic Acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.