accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CSOH2N

CAS RN 957344-92-2

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxamide
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxamide
Molecular Weight (g/mol): 253.12
Molecular Formula: C11H16BNO3S
InChi Key: SQHNPVGOAXWLAM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(S1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results