Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Carbamoylthiophene-2-boronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH5235303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5235303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5235303 Supplier Thermo Scientific Chemicals Supplier No. H5235303

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Carbamoylthiophene-2-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957344-92-2
Molecular Formula C11H16BNO3S
Molecular Weight (g/mol) 253.12
MDL Number MFCD17215214
InChI Key SQHNPVGOAXWLAM-UHFFFAOYSA-N
Synonym 5-carbamoylthiophene-2-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl thiophene-2-carboxamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophene-2-carboxamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxa-borolan-2-yl-thiophene-2-carboxylic acid amide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-thiophene-2-carboxylic acid amide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl thiophene-2-carboxylic acid amide
PubChem CID 57589940
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carboxamide
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(S1)C(N)=O

Specifications

Quantity 1 g
Formula Weight 253.13
Percent Purity 96%
Chemical Name or Material 5-Carbamoylthiophene-2-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.