accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 957780-54-0

OOOOHOOOONNHHN(S)(S)H3CH3CH3CH3CCH3

CAS RN 957780-54-0

IUPAC Name: (4S)-3-[(2S)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Synonyms: (4S)-3-[(2S)-6-[(tert-butoxycarbonyl)amino]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
Molecular Weight (g/mol): 595.69
Molecular Formula: C32H41N3O8
InChi Key: SCQJDNYUMSKATK-UIOOFZCWSA-N
SMILES: CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(=O)N1[C@@H](COC1(C)C)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)