CAS RN 958032-89-8
CAS RN 958032-89-8
IUPAC Name:
1-(but-3-en-1-yloxy)-2,3,3,4,4,5,5-heptafluorocyclopent-1-ene
Synonyms:
1-(but-3-en-1-yloxy)-2,3,3,4,4,5,5-heptafluorocyclopent-1-ene
Molecular Weight (g/mol):
264.14
Molecular Formula:
C9H7F7O
InChi Key:
LEWPTLXFUJDZPB-UHFFFAOYSA-N
SMILES:
FC1=C(OCCC=C)C(F)(F)C(F)(F)C1(F)F
1
–
1
of
1
results
1
1-(3-Butenyloxy)-2,3,3,4,4,5,5-heptafluorocyclopentene 97.0+%, TCI America™
CAS: 958032-89-8 Molecular Formula: C9H7F7O Molecular Weight (g/mol): 264.143 MDL Number: MFCD00084878 InChI Key: LEWPTLXFUJDZPB-UHFFFAOYSA-N Synonym: BHFC PubChem CID: 59259641 IUPAC Name: 1-but-3-enoxy-2,3,3,4,4,5,5-heptafluorocyclopentene SMILES: C=CCCOC1=C(C(C(C1(F)F)(F)F)(F)F)F